Search
Search

Unc5d| Gene Symbol: Unc5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153135.3 | NP_694775.1 | 14 - 15 | 2468 | 69 | |
| GenBank mRNA | AJ487854.1 | - | 14 - 15 | 2469 | 69 | |
| AK220417.1 | - | 4 - 5 | 809 | 69 | ||
| BC137615.1 | - | 14 - 15 | 2259 | 8860634 | 69 | |
| BC144783.1 | - | 12 - 13 | 2076 | 9053331 | 69 | |
| BC144784.1 | - | 12 - 13 | 2037 | 9053332 | 69 | |
| BC145695.1 | - | 11 - 12 | 2043 | 40130998 | 69 | |
| FJ824810.1 | - | 13 - 14 | 2347 | 69 | ||
Set Membership: |
|