Search
Search

Cdc16| Gene Symbol: Cdc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027276.2 | NP_081552.2 | 9 - 10 | 963 | 95 | |
| GenBank mRNA | AK147938.1 | - | 9 - 10 | 978 | 95 | |
| AK167965.1 | - | 9 - 10 | 978 | 95 | ||
| AK168978.1 | - | 9 - 10 | 950 | 95 | ||
| AK189595.1 | - | 4 - 5 | 395 | 95 | ||
| AK213896.1 | - | 2 - 3 | 151 | 95 | ||
| BC026606.1 | - | 9 - 10 | 963 | 4976424 | 95 | |
Set Membership: |
|