Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01220741_m1
          See other A630089N07RIK GE Assays ›
          Gene Symbol:
          A630089N07Rik
          Gene Name:
          RIKEN cDNA A630089N07 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 98062512 - 98082439 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01220741_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A630089N07Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015491.1 4 - 5 401 89
          GenBank mRNA AK041610.1 - 4 - 5 413 89
          AK042413.1 - 4 - 5 402 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600002162.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017924 AK042413.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017924 NR_015491.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P205812 BC027541.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P31002531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31002487 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31002495 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31002504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31002477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31002506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31002474 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10532312 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532306 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532307 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532311 - -
          Affymetrix Mouse Gene 1.1 ST Array 10441339 - -
          Affymetrix Mouse Clariom S Array PSR1600016772.mm.1 NR_015491.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600016772.mm.1 NR_015491.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600016772.mm.1 TC1600002162.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600016773.mm.1 NR_015491.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600016773.mm.1 NR_015491.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600016773.mm.1 TC1600002162.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600016789.mm.1 NR_015491.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600016789.mm.1 NR_015491.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600016789.mm.1 TC1600002162.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600016780.mm.1 NR_015491.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600016780.mm.1 NR_015491.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600016780.mm.1 TC1600002162.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600016781.mm.1 NR_015491.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600016781.mm.1 NR_015491.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600016781.mm.1 TC1600002162.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600016776.mm.1 NR_015491.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600016776.mm.1 NR_015491.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600016776.mm.1 TC1600002162.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline