Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01221823_s1
          See other 4930480E11RIK GE Assays ›
          Gene Symbol:
          4930480E11Rik
          Gene Name:
          RIKEN cDNA 4930480E11 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 78369643 - 78371128 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01221823_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930480E11Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177966.2 NP_001171437.1 1 1354 142

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156302 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043582 BC116861.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043582 BC116859.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043582 NM_001177966.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043582 AK015598.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156302 - -
          Whole Mouse Genome G4122A A_51_P364550 AK015598.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156302 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14778601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14818431 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15063086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14818432 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15066390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15066388 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481011 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15066389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481042 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32480999 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10600582 - -
          Affymetrix HT MG-430 PM Array Plate 1454100_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10600581 - -
          Mouse Genome 430 2.0 Array 1454100_PM_at - -
          Mouse Genome 430 2.0 Array 1454100_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74910_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74910_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00005308.mm.1 NM_001177966.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005308.mm.1 TC0X00000784.mm.1 MTA TC
          Mouse Expression Set 430 1454100_PM_at - -
          Mouse Expression Set 430 1454100_at - -
          Affymetrix HT Mouse Genome 430B Array 1454100_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454100_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 385926 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258502 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2721584 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline