Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mm01221912_m1
See other ERCC6 GE Assays ›
Gene Symbol:
Ercc6
Gene Name:
excision repair cross-complementing rodent repair deficiency, complementation group 6
Gene Aliases:
4732403I04, C130058G22Rik, CSB
Chromosome Location:
Chr.14: 32513288 - 32580990 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID
Mm01221912_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Ercc6
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001081221.1
NP_001074690.1
10 - 11
2244
97
GenBank mRNA
BC132447.1
-
6 - 7
1183
97
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as MTA transcript cluster(s) TC1400000347.mm.1
Set Membership:
Amplicon length between 86 and 100
Mammalian Gene Collection
Mus musculus
Probe spans exons
Gene Ontology Categories:
Function(s)
Process(es)
response to superoxide
DNA repair
transcription-coupled nucleotide-excision repair
base-excision repair
pyrimidine dimer repair
transcription elongation from RNA polymerase I promoter
cellular response to DNA damage stimulus
response to oxidative stress
activation of JNKK activity
activation of JUN kinase activity
intrinsic apoptotic signaling pathway in response to DNA damage
response to UV
response to toxic substance
response to X-ray
response to UV-B
response to gamma radiation
regulation of DNA-templated transcription, elongation
positive regulation of DNA-templated transcription, elongation
multicellular organism growth
photoreceptor cell maintenance
positive regulation of protein tyrosine kinase activity
DNA binding
chromatin binding
ATP binding
protein C-terminus binding
DNA-dependent ATPase activity
protein tyrosine kinase activator activity
protein complex binding
protein N-terminus binding
MicroArray Probes:
Affymetrix
Array Name
Probe IDs
Targets
Target Type
Affymetrix Mouse Clariom S Array
PSR1400003074.mm.1
NM_001081221.1.1
Refseq
Affymetrix Mouse Clariom S Array
PSR1400003074.mm.1
NM_001081221.1
Refseq
Affymetrix Mouse Clariom S Array
PSR1400003074.mm.1
TC1400000347.mm.1
MTA TC
Affymetrix Mouse Clariom S Array
PSR1400003075.mm.1
NM_001081221.1.1
Refseq
Affymetrix Mouse Clariom S Array
PSR1400003075.mm.1
NM_001081221.1
Refseq
Affymetrix Mouse Clariom S Array
PSR1400003075.mm.1
TC1400000347.mm.1
MTA TC
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline