Search
Search

Ogdhl| Gene Symbol: Ogdhl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081130.1 | NP_001074599.1 | 19 - 20 | 2612 | 72 | |
| GenBank mRNA | BC157970.1 | - | 18 - 19 | 2412 | 9007292 | 72 |
| BC157971.1 | - | 18 - 19 | 2412 | 9007293 | 72 | |
Set Membership: |
|