Search
Search
| Gene Symbol: Gab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181584.4 | NP_853615.1 | 2 - 3 | 537 | 80 | |
| GenBank mRNA | AK031227.1 | - | 2 - 3 | 535 | 80 | |
| AY057988.1 | - | 2 - 3 | 423 | 80 | ||
| BC141240.1 | - | 2 - 3 | 417 | 9055852 | 80 | |
Set Membership: |
|