Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01222137_g1
          See other MPEG1 GE Assays ›
          Gene Symbol:
          Mpeg1
          Gene Name:
          macrophage expressed gene 1
          Gene Aliases:
          MPS1, Mpg-1
          Chromosome Location:
          Chr.19: 12460709 - 12465285 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01222137_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mpeg1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010821.1 NP_034951.1 1 - 2 81 67
          GenBank mRNA AK030216.1 - 1 - 2 82 67

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the NM_010821.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P180373 BC129844.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P180373 L20315.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P180373 BC049256.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P180373 BC129843.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P180373 AK030216.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P180373 BC129844.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P180373 L20315.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P180373 BC049256.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P180373 BC129843.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P180373 AK030216.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P390538 L20315.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P390538 BC049256.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P390538 AK149517.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P390538 NM_010821.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P390538 L20315.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P390538 BC049256.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P390538 AK149517.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P390538 NM_010821.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 681925 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 838791 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P178543 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P390538 L20315.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P390538 BC049256.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P390538 AK149517.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P390538 NM_010821.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P390538 L20315.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P390538 BC049256.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P390538 AK149517.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P390538 NM_010821.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P180373 BC129844.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P180373 L20315.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P180373 BC049256.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P180373 BC129843.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P180373 AK030216.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P180373 BC129844.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P180373 L20315.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P180373 BC049256.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P180373 BC129843.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P180373 AK030216.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P178543 - -
          Whole Mouse Genome G4122A A_51_P390538 L20315.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P390538 BC049256.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P390538 AK149517.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P390538 NM_010821.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P390538 L20315.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P390538 BC049256.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P390538 AK149517.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P390538 NM_010821.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P180373 BC129844.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P180373 L20315.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P180373 BC049256.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P180373 BC129843.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P180373 AK030216.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P180373 BC129844.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P180373 L20315.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P180373 BC049256.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P180373 BC129843.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P180373 AK030216.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132390 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P178543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14214108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960813 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14304417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14414675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14233549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14304424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14304460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14363679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14304411 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960805 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14304420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960877 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461722 - -
          Affymetrix HT MG-430 PM Array Plate 1427076_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10461721 - -
          Mouse Genome 430 2.0 Array 1427076_PM_at - -
          Mouse Genome 430 2.0 Array 1427076_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17476_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17476_at - -
          Affymetrix Murine Genome 6500A Array Msa16000_at - -
          Affymetrix Mouse Clariom S Array PSR1900002903.mm.1 NM_010821.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002903.mm.1 NM_010821.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002903.mm.1 TC1900000246.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002906.mm.1 NM_010821.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002906.mm.1 NM_010821.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002906.mm.1 TC1900000246.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002905.mm.1 NM_010821.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002905.mm.1 NM_010821.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002905.mm.1 TC1900000246.mm.1 MTA TC
          Mouse Expression Set 430 1427076_PM_at - -
          Mouse Expression Set 430 1427076_at - -
          Murine Genome U74v2 Set 162075_r_at - -
          Murine Genome U74v2 Set 99071_PM_at - -
          Murine Genome U74v2 Set 99071_at - -
          Murine Genome U74v2 Set 162075_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1427076_at - -
          Mouse Genome 430A 2.0 Array 1427076_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427076_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427076_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 551683 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103780671 - -
          MouseWG-6 v2 Expression BeadChip 105220471 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247377 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline