Search
Search
| Gene Symbol: Nek3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162947.1 | NP_001156419.1 | 2 - 3 | 259 | 76 | |
| NM_011848.4 | NP_035978.2 | 3 - 4 | 317 | 76 | ||
| GenBank mRNA | AF093416.1 | - | 2 - 3 | 264 | 76 | |
| AF099066.1 | - | 3 - 4 | 286 | 76 | ||
| BC005411.1 | - | 3 - 4 | 294 | 3590866 | 76 | |
| BU058938.1 | - | 3 - 4 | 317 | 76 | ||
Set Membership: |
|