Search
Search

Clock| Gene Symbol: Clock | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289826.1 | NP_001276755.1 | 8 - 9 | 779 | 132 | |
| NM_007715.6 | NP_031741.1 | 8 - 9 | 822 | 132 | ||
| GenBank mRNA | AB221635.1 | - | 5 - 6 | 434 | 132 | |
| AF000998.1 | - | 8 - 9 | 822 | 132 | ||
| AK040337.1 | - | 8 - 9 | 760 | 132 | ||
| AK129118.1 | - | 8 - 9 | 779 | 132 | ||
| BC129886.1 | - | 6 - 7 | 531 | 132 | ||
Set Membership: |
|