Search
Search

Lmbr1| Gene Symbol: Lmbr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020295.3 | NP_064691.2 | 14 - 15 | 1360 | 76 | |
| GenBank mRNA | AF190665.1 | - | 14 - 15 | 1162 | 76 | |
| AK030631.1 | - | 15 - 16 | 2166 | 76 | ||
| BC016110.1 | - | 14 - 15 | 1119 | 4481088 | 76 | |
Set Membership: |
|