Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01222895_gH
          See other DKC1 GE Assays ›
          Gene Symbol:
          Dkc1
          Gene Name:
          dyskeratosis congenita 1, dyskerin
          Gene Aliases:
          BC068171
          Chromosome Location:
          Chr.X: 75095854 - 75109777 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01222895_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dkc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001030307.2 NP_001025478.1 5 - 6 705 64
          GenBank mRNA AK136418.1 - 5 - 6 490 64
          AK144139.1 - 5 - 6 493 64
          AK166105.1 - 5 - 6 481 64
          AK167973.1 - 5 - 6 456 64
          BC068171.1 - 5 - 6 461 64

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Panther Classification:

          Molecular Function -

          centromere DNA-binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          rRNA modification
          box H/ACA snoRNA 3'-end processing
          pseudouridine synthesis
          RNA-dependent DNA biosynthetic process
          rRNA processing
          RNA processing
          cell proliferation
          RNA modification
          rRNA pseudouridine synthesis
          snRNA pseudouridine synthesis
          positive regulation of telomere maintenance via telomerase
          box H/ACA snoRNA metabolic process
          ribosome biogenesis
          positive regulation of telomerase activity
          DNA biosynthetic process
          mRNA pseudouridine synthesis
          telomerase activity
          RNA binding
          pseudouridine synthase activity
          isomerase activity
          pivalyl-CoA mutase activity
          o-hydroxylaminobenzoate mutase activity
          lupeol synthase activity
          beta-amyrin synthase activity
          poly(A) RNA binding
          baruol synthase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0X00005095.mm.1 NM_001030307.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005095.mm.1 NM_001030307.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005095.mm.1 TC0X00000735.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005090.mm.1 NM_001030307.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005090.mm.1 NM_001030307.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005090.mm.1 TC0X00000735.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005088.mm.1 NM_001030307.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005088.mm.1 NM_001030307.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005088.mm.1 TC0X00000735.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005094.mm.1 NM_001030307.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005094.mm.1 NM_001030307.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005094.mm.1 TC0X00000735.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005093.mm.1 NM_001030307.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005093.mm.1 NM_001030307.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005093.mm.1 TC0X00000735.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005085.mm.1 NM_001030307.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005085.mm.1 NM_001030307.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005085.mm.1 TC0X00000735.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline