Search
Search

Ccnl1| Gene Symbol: Ccnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019937.3 | NP_064321.2 | 6 - 7 | 888 | 84 | |
| GenBank mRNA | AF159159.1 | - | 6 - 7 | 841 | 84 | |
| AF467251.1 | - | 6 - 7 | 882 | 84 | ||
| AK011629.1 | - | 6 - 7 | 845 | 84 | ||
| BC007177.1 | - | 6 - 7 | 2624 | 84 | ||
| BC037204.1 | - | 5 - 6 | 2515 | 84 | ||
| BC069909.1 | - | 7 - 8 | 972 | 84 | ||
| BC094383.1 | - | 6 - 7 | 824 | 30604933 | 84 | |
| BC117816.1 | - | 7 - 8 | 781 | 84 | ||
Set Membership: |
|