Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01223097_m1
          See other CDK8 GE Assays ›
          Gene Symbol:
          Cdk8
          Gene Name:
          cyclin-dependent kinase 8
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 146230736 - 146302874 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01223097_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdk8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153599.3 NP_705827.2 6 - 7 665 78
          GenBank mRNA CF723869.1 - 4 - 5 367 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001779.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdk8

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein phosphorylation
          phosphorylation
          protein ubiquitination
          negative regulation of triglyceride metabolic process
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          cyclin-dependent protein serine/threonine kinase activity
          ATP binding
          RNA polymerase II carboxy-terminal domain kinase activity
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          ubiquitin protein ligase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500016475.mm.1 TC0500001779.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500016475.mm.1 NM_153599.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016475.mm.1 NM_153599.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016492.mm.1 TC0500001779.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500016492.mm.1 NM_153599.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016492.mm.1 NM_153599.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016487.mm.1 TC0500001779.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500016487.mm.1 NM_153599.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016487.mm.1 NM_153599.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016485.mm.1 TC0500001779.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500016485.mm.1 NM_153599.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016485.mm.1 NM_153599.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016483.mm.1 TC0500001779.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500016483.mm.1 NM_153599.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016483.mm.1 NM_153599.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016478.mm.1 TC0500001779.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500016478.mm.1 NM_153599.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016478.mm.1 NM_153599.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016484.mm.1 TC0500001779.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500016484.mm.1 NM_153599.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016484.mm.1 NM_153599.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016481.mm.1 TC0500001779.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500016481.mm.1 NM_153599.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016481.mm.1 NM_153599.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016482.mm.1 TC0500001779.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500016482.mm.1 NM_153599.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016482.mm.1 NM_153599.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016486.mm.1 TC0500001779.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500016486.mm.1 NM_153599.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016486.mm.1 NM_153599.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline