Search
Search

Aftph| Gene Symbol: Aftph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252503.2 | NP_001239432.1 | 6 - 7 | 2664 | 107 | |
| NM_001290545.1 | NP_001277474.1 | 6 - 7 | 2664 | 107 | ||
| NM_181411.4 | NP_852076.1 | 6 - 7 | 2664 | 107 | ||
| GenBank mRNA | AK029681.1 | - | 6 - 7 | 2564 | 107 | |
| AK030572.1 | - | 6 - 7 | 2608 | 107 | ||
| AK047225.1 | - | 5 - 6 | 1728 | 107 | ||
| AK136815.1 | - | 5 - 6 | 1916 | 107 | ||
| AK150458.1 | - | 4 - 5 | 455 | 107 | ||
| AK152308.1 | - | 4 - 5 | 455 | 107 | ||
| AK152487.1 | - | 4 - 5 | 455 | 107 | ||
| AK161532.1 | - | 4 - 5 | 468 | 107 | ||
| BC004630.1 | - | 5 - 6 | 953 | 107 | ||
| BC039995.1 | - | 1 - 2 | 122 | 107 | ||
| BC052036.1 | - | 6 - 7 | 2592 | 5702534 | 107 | |
Set Membership: |
|