Search
Search

Atp6v1h| Gene Symbol: Atp6v1h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133826.4 | NP_598587.2 | 9 - 10 | 1005 | 135 | |
| GenBank mRNA | AK034165.1 | - | 7 - 8 | 3164 | 135 | |
| AK078767.1 | - | 9 - 10 | 1006 | 135 | ||
| AK152147.1 | - | 9 - 10 | 1027 | 135 | ||
| BC009154.1 | - | 9 - 10 | 1006 | 3601621 | 135 | |
Set Membership: |
|