Search
Search

Arhgap28| Gene Symbol: Arhgap28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172964.4 | NP_766552.3 | 2 - 3 | 436 | 152 | |
| GenBank mRNA | AK087523.1 | - | 2 - 3 | 449 | 152 | |
| AK133720.1 | - | 2 - 3 | 278 | 152 | ||
| AK141901.1 | - | 2 - 3 | 386 | 152 | ||
| BC066788.1 | - | 2 - 3 | 426 | 30049271 | 152 | |
Set Membership: |
|