Search
Search

Tmtc4| Gene Symbol: Tmtc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028651.2 | NP_082927.1 | 2 - 3 | 303 | 100 | |
| GenBank mRNA | AK028333.1 | - | 2 - 3 | 284 | 100 | |
| AK033947.1 | - | 2 - 3 | 304 | 100 | ||
| AK036741.1 | - | 2 - 3 | 260 | 100 | ||
| AK039093.1 | - | 2 - 3 | 248 | 100 | ||
| AK043293.1 | - | 2 - 3 | 330 | 100 | ||
| AK044391.1 | - | 2 - 3 | 259 | 100 | ||
| AK045271.1 | - | 2 - 3 | 265 | 100 | ||
| AK049748.1 | - | 3 - 4 | 444 | 100 | ||
| AK082526.1 | - | 1 - 2 | 131 | 100 | ||
Set Membership: |
|