Search
Search

Ddx11| Gene Symbol: Ddx11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003919.1 | NP_001003919.1 | 14 - 15 | 1574 | 61 | |
| GenBank mRNA | AK049419.1 | - | 14 - 15 | 1580 | 61 | |
| AK076357.1 | - | 15 - 16 | 1604 | 61 | ||
| AK152868.1 | - | 14 - 15 | 1562 | 61 | ||
| BC049929.1 | - | 2 - 3 | 88 | 61 | ||
| BC079656.1 | - | 14 - 15 | 1574 | 6854039 | 61 | |
Set Membership: |
|