Search
Search

Ptprr| Gene Symbol: Ptprr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161837.1 | NP_001155309.1 | 10 - 11 | 1398 | 81 | |
| NM_001161838.1 | NP_001155310.1 | 7 - 8 | 1355 | 81 | ||
| NM_001161839.1 | NP_001155311.1 | 8 - 9 | 1046 | 81 | ||
| NM_001161840.1 | NP_001155312.1 | 8 - 9 | 1238 | 81 | ||
| NM_011217.2 | NP_035347.1 | 11 - 12 | 2004 | 81 | ||
| GenBank mRNA | AF041866.1 | - | 8 - 9 | 1163 | 81 | |
| AF129509.1 | - | 7 - 8 | 1280 | 81 | ||
| AK028054.1 | - | 11 - 12 | 2005 | 81 | ||
| BC119230.1 | - | 11 - 12 | 1937 | 8733979 | 81 | |
| BC120547.1 | - | 11 - 12 | 1937 | 40129470 | 81 | |
| D31898.1 | - | 11 - 12 | 1967 | 81 | ||
| Z30313.1 | - | 10 - 11 | 1393 | 81 | ||
Set Membership: |
|