Search
Search
| Gene Symbol: Kif4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008446.2 | NP_032472.2 | 19 - 20 | 2644 | 71 | |
| GenBank mRNA | AK135430.1 | - | 19 - 20 | 2138 | 71 | |
| AK171938.1 | - | 19 - 20 | 2556 | 71 | ||
| BC050946.1 | - | 19 - 20 | 2582 | 6822283 | 71 | |
| BC054537.1 | - | 19 - 20 | 2566 | 71 | ||
| D12646.1 | - | 19 - 20 | 2971 | 71 | ||
Set Membership: |
|