Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01228041_g1
          See other AMT GE Assays ›
          Gene Symbol:
          Amt
          Gene Name:
          aminomethyltransferase
          Gene Aliases:
          EG434437
          Chromosome Location:
          Chr.9: 108296922 - 108301722 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01228041_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Amt
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013814.1 NP_001013836.1 8 - 9 1048 103
          GenBank mRNA BC132277.1 - 8 - 9 1100 40130554 103
          BC132279.1 - 8 - 9 1100 40130556 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001399.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Amt

          Panther Classification:

          Molecular Function -

          methyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          glycine catabolic process
          aminomethyltransferase activity
          transaminase activity
          transferase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1975052 NM_001013814.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914978 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 2400 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 736086 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1975052 NM_001013814.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814162 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06919946 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07907256 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07907261 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07907260 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07907265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07964226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07964227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07907261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07907266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07659984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07613160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07907254 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10588885 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588891 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588884 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588889 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588886 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588888 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588890 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588887 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588892 - -
          Affymetrix Mouse Gene 1.1 ST Array 10588883 - -
          Affymetrix Mouse Clariom S Array PSR0900011234.mm.1 NM_001013814.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011234.mm.1 NM_001013814.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011234.mm.1 TC0900001399.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900011233.mm.1 NM_001013814.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011233.mm.1 NM_001013814.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011233.mm.1 TC0900001399.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900011231.mm.1 NM_001013814.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011231.mm.1 NM_001013814.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011231.mm.1 TC0900001399.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160992 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160992 - -
          mouse-6 v1.1 (Illumina) GI_62000669-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160992 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160992 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline