Search
Search

Bfsp2| Gene Symbol: Bfsp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002896.2 | NP_001002896.1 | 3 - 4 | 828 | 95 | |
| GenBank mRNA | AK134810.1 | - | 3 - 4 | 760 | 95 | |
| AK193757.1 | - | 1 - 2 | 119 | 95 | ||
| BC068172.1 | - | 3 - 4 | 828 | 30550909 | 95 | |
| S55549.1 | - | 1 - 2 | 137 | 95 | ||
Set Membership: |
|