Search
Search
| Gene Symbol: Trf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133977.2 | NP_598738.1 | 9 - 10 | 1273 | 72 | |
| GenBank mRNA | AF440692.1 | - | 8 - 9 | 1203 | 72 | |
| AK085754.1 | - | 9 - 10 | 1258 | 72 | ||
| AK142599.1 | - | 1263 | 72 | |||
| AK146549.1 | - | 9 - 10 | 1258 | 72 | ||
| AK149559.1 | - | 9 - 10 | 1258 | 72 | ||
| AK149595.1 | - | 9 - 10 | 1258 | 72 | ||
| AK150782.1 | - | 9 - 10 | 1306 | 72 | ||
| AK168368.1 | - | 9 - 10 | 1260 | 72 | ||
| AK168405.1 | - | 9 - 10 | 1260 | 72 | ||
| AK168419.1 | - | 9 - 10 | 1258 | 72 | ||
| AK168938.1 | - | 9 - 10 | 1258 | 72 | ||
| AK169187.1 | - | 9 - 10 | 1257 | 72 | ||
| BC008559.1 | - | 7 - 8 | 995 | 72 | ||
| BC012313.1 | - | 9 - 10 | 1273 | 3981648 | 72 | |
| BC018573.1 | - | 4 - 5 | 543 | 72 | ||
| BC020295.1 | - | 9 - 10 | 1242 | 72 | ||
| BC022986.1 | - | 9 - 10 | 1251 | 5123741 | 72 | |
| BC058216.1 | - | 9 - 10 | 1239 | 72 | ||
| BC058218.1 | - | 9 - 10 | 1252 | 72 | ||
| BC092046.1 | - | 9 - 10 | 1234 | 3594705 | 72 | |
Set Membership: |
|