Search
Search

Traip| Gene Symbol: Traip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011634.3 | NP_035764.2 | 8 - 9 | 816 | 85 | |
| GenBank mRNA | AK012786.1 | - | 8 - 9 | 798 | 85 | |
| AK012948.1 | - | 8 - 9 | 815 | 85 | ||
| U77844.1 | - | 8 - 9 | 821 | 85 | ||
Set Membership: |
|