Search
Search
| Gene Symbol: Apeh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146226.2 | NP_666338.2 | 19 - 20 | 1918 | 81 | |
| GenBank mRNA | AK149671.1 | - | 19 - 20 | 1924 | 81 | |
| AK149713.1 | - | 19 - 20 | 1920 | 81 | ||
| AK178617.1 | - | 1 - 2 | 147 | 81 | ||
| BC005716.1 | - | 18 - 19 | 2044 | 81 | ||
| BC023937.1 | - | 9 - 10 | 887 | 81 | ||
| BC025494.1 | - | 19 - 20 | 1855 | 5310043 | 81 | |
| BC026594.1 | - | 14 - 15 | 1451 | 81 | ||
| BC034199.1 | - | 4 - 5 | 421 | 81 | ||
Set Membership: |
|