Search
Search

Itm2a| Gene Symbol: Itm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008409.2 | NP_032435.2 | 3 - 4 | 580 | 64 | |
| GenBank mRNA | AK014024.1 | - | 3 - 4 | 348 | 64 | |
| AK051956.1 | - | 3 - 4 | 559 | 64 | ||
| AK135229.1 | - | 3 - 4 | 532 | 64 | ||
| AK167351.1 | - | 3 - 4 | 581 | 64 | ||
| BC031420.1 | - | 3 - 4 | 547 | 3668557 | 64 | |
| L38971.1 | - | 3 - 4 | 583 | 64 | ||
Set Membership: |
|