Search
Search

Bscl2| Gene Symbol: Bscl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136064.3 | NP_001129536.1 | 9 - 10 | 1593 | 74 | |
| NM_001290823.1 | NP_001277752.1 | 8 - 9 | 1052 | 74 | ||
| NM_008144.5 | NP_032170.3 | 8 - 9 | 1443 | 74 | ||
| GenBank mRNA | AB030196.1 | - | 8 - 9 | 1062 | 74 | |
| AF069954.2 | - | 8 - 9 | 1268 | 74 | ||
| AK004634.1 | - | 6 - 7 | 3142 | 74 | ||
| AK151606.1 | - | 8 - 9 | 1021 | 74 | ||
| AK155753.1 | - | 7 - 8 | 1560 | 74 | ||
| AK158901.1 | - | 8 - 9 | 1444 | 74 | ||
| AK170183.1 | - | 8 - 9 | 1019 | 74 | ||
| BC043023.1 | - | 8 - 9 | 1207 | 5689013 | 74 | |
| BC061689.1 | - | 9 - 10 | 1331 | 6306706 | 74 | |
Set Membership: |
|