Search
Search

Slc3a2| Gene Symbol: Slc3a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161413.1 | NP_001154885.1 | 6 - 7 | 1190 | 74 | |
| NM_008577.4 | NP_032603.3 | 5 - 6 | 949 | 74 | ||
| GenBank mRNA | AB023408.1 | - | 5 - 6 | 904 | 74 | |
| AK090046.1 | - | 5 - 6 | 927 | 74 | ||
| AK156285.1 | - | 5 - 6 | 421 | 74 | ||
| AK161280.1 | - | 6 - 7 | 1191 | 74 | ||
| AK165417.1 | - | 5 - 6 | 931 | 74 | ||
| AK165550.1 | - | 5 - 6 | 927 | 74 | ||
| AK183775.1 | - | 4 - 5 | 314 | 74 | ||
| AK184227.1 | - | 2 - 3 | 76 | 74 | ||
| AK198833.1 | - | 170 | 74 | |||
| AK215848.1 | - | 3 - 4 | 174 | 74 | ||
| BC065173.1 | - | 5 - 6 | 909 | 6415269 | 74 | |
| X14309.1 | - | 5 - 6 | 891 | 74 | ||
Set Membership: |
|