Search
Search
| Gene Symbol: Mtl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039657.2 | NP_001034746.1 | 4 - 5 | 830 | 74 | |
| NM_001039658.2 | NP_001034747.1 | 4 - 5 | 830 | 74 | ||
| NM_001286557.1 | NP_001273486.1 | 5 - 6 | 914 | 74 | ||
| NM_001286558.1 | NP_001273487.1 | 4 - 5 | 845 | 74 | ||
| GenBank mRNA | AB057423.1 | - | 4 - 5 | 818 | 74 | |
| AF329359.1 | - | 4 - 5 | 855 | 74 | ||
| AF329360.1 | - | 5 - 6 | 924 | 74 | ||
| AK015732.1 | - | 4 - 5 | 803 | 74 | ||
| AK161511.1 | - | 4 - 5 | 831 | 74 | ||
| BC024377.1 | - | 4 - 5 | 806 | 4976492 | 74 | |
| U67176.1 | - | 6 - 7 | 847 | 74 | ||
| U77383.1 | - | 4 - 5 | 467 | 74 | ||
Set Membership: |
|