Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01231228_g1
          See other NUDT8 GE Assays ›
          Gene Symbol:
          Nudt8
          Gene Name:
          nudix (nucleoside diphosphate linked moiety X)-type motif 8
          Gene Aliases:
          2310039H17Rik
          Chromosome Location:
          Chr.19: 4000580 - 4002102 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01231228_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nudt8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025529.3 NP_079805.1 3 - 4 474 66
          GenBank mRNA AK002605.1 - 3 - 4 412 66
          AK009700.1 - 3 - 4 412 66
          AK045197.1 - 3 - 4 475 66
          AK170076.1 - 3 - 4 414 66
          BC056443.1 - 3 - 4 414 6822891 66

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nudt8

          Panther Classification:

          Molecular Function -

          hydrolase phosphatase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          thiamine-pyrophosphatase activity
          8-oxo-7,8-dihydroguanosine triphosphate pyrophosphatase activity
          UDP-2,3-diacylglucosamine hydrolase activity
          bis(5'-nucleosyl)-tetraphosphatase activity
          dATP pyrophosphohydrolase activity
          pyrophosphatase activity
          hydrolase activity
          dihydroneopterin monophosphate phosphatase activity
          dihydroneopterin triphosphate pyrophosphohydrolase activity
          dITP diphosphatase activity
          dTTP diphosphatase activity
          XTP diphosphatase activity
          ATP-dependent 5'-3' DNA helicase activity
          phosphocholine hydrolase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31920557 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31920556 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31920558 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P490388 NM_025529.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P490388 AK002605.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P490388 AK009700.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P490388 AK045197.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P490388 AK170076.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P490388 NM_025529.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P490388 AK002605.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P490388 AK009700.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P490388 AK045197.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P490388 AK170076.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 167453 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102869 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P490388 NM_025529.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P490388 AK002605.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P490388 AK009700.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P490388 AK045197.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P490388 AK170076.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P490388 NM_025529.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P490388 AK002605.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P490388 AK009700.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P490388 AK045197.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P490388 AK170076.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P490388 NM_025529.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P490388 AK002605.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P490388 AK009700.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P490388 AK045197.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P490388 AK170076.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P490388 NM_025529.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P490388 AK002605.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P490388 AK009700.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P490388 AK045197.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P490388 AK170076.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31920556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31920557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31920558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31920562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31920564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31920567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31920566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31920563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31920561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31920569 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102869 - -
          Whole Mouse Genome G4122A A_51_P490388 NM_025529.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P490388 AK002605.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P490388 AK009700.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P490388 AK045197.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P490388 AK170076.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P490388 NM_025529.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P490388 AK002605.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P490388 AK009700.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P490388 AK045197.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P490388 AK170076.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136990 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102869 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14151807 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14151825 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14151854 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14151810 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14151848 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14151853 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14151852 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14151834 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31920556 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31920558 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31920557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31920564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31920569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31920553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14151815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31920550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31920549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31920561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31920551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31920558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31920554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31920566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31920552 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10460286 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460288 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460289 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460287 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460290 - -
          Affymetrix HT MG-430 PM Array Plate 1450111_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10460285 - -
          Mouse Genome 430 2.0 Array 1450111_PM_a_at - -
          Mouse Genome 430 2.0 Array 1450111_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66387_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66387_at - -
          Mouse Expression Set 430 1450111_a_at - -
          Mouse Expression Set 430 1450111_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1450111_a_at - -
          Mouse Genome 430A 2.0 Array 1450111_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1450111_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1450111_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3710441 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003710441 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2617305 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2617305 - -
          mouse-6 v1.1 (Illumina) scl52798.4.1_101-S - -
          Illumina Sentrix mouse-6.v1 bead array 3710441 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52798.4.1_101-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3710441 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2617305 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2617305 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline