Search
Search

Pcnxl3| Gene Symbol: Pcnxl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144868.3 | NP_659117.2 | 32 - 33 | 6212 | 80 | |
| GenBank mRNA | AF237953.1 | - | 30 - 31 | 4229 | 80 | |
| AK144114.1 | - | 8 - 9 | 1472 | 80 | ||
| AK220471.1 | - | 10 - 11 | 2110 | 80 | ||
| BC038056.1 | - | 27 - 28 | 4320 | 80 | ||
| BC054751.1 | - | 32 - 33 | 5491 | 80 | ||
| BC055289.1 | - | 33 - 34 | 6015 | 80 | ||
| BC096538.1 | - | 32 - 33 | 6212 | 80 | ||
Set Membership: |
|