Search
Search

Snx32| Gene Symbol: Snx32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024560.2 | NP_001019731.1 | 8 - 9 | 908 | 75 | |
| GenBank mRNA | AK047234.1 | - | 8 - 9 | 884 | 75 | |
| BC048922.1 | - | 8 - 9 | 908 | 6311864 | 75 | |
Set Membership: |
|