Search
Search

Macrod1| Gene Symbol: Macrod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134147.4 | NP_598908.2 | 9 - 10 | 1092 | 58 | |
| GenBank mRNA | AI604841.1 | - | 2 - 3 | 58 | ||
| AK052998.1 | - | 9 - 10 | 1051 | 58 | ||
Set Membership: |
|