Search
Search

Lpin2| Gene Symbol: Lpin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164885.1 | NP_001158357.1 | 3 - 4 | 523 | 115 | |
| NM_022882.4 | NP_075020.2 | 3 - 4 | 399 | 115 | ||
| GenBank mRNA | AF286723.1 | - | 3 - 4 | 399 | 115 | |
| AK033662.1 | - | 3 - 4 | 436 | 115 | ||
| AK050018.1 | - | 3 - 4 | 524 | 115 | ||
| AK086834.1 | - | 3 - 4 | 335 | 115 | ||
| AK136472.1 | - | 994 | 115 | |||
| BC039698.1 | - | 3 - 4 | 512 | 5101211 | 115 | |
Set Membership: |
|