Search
Search

Klhdc8b| Gene Symbol: Klhdc8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030075.2 | NP_084351.1 | 6 - 7 | 1094 | 57 | |
| GenBank mRNA | AK019849.1 | - | 6 - 7 | 1092 | 57 | |
| AK154428.1 | - | 6 - 7 | 1195 | 57 | ||
| BC048918.1 | - | 1 - 2 | 57 | 6309434 | 57 | |
| BC060089.1 | - | 1 - 2 | 398 | 57 | ||
| CA750541.1 | - | 1 - 2 | 337 | 57 | ||
Set Membership: |
|