Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01232629_g1
          See other COL6A6 GE Assays ›
          Gene Symbol:
          Col6a6
          Gene Name:
          collagen, type VI, alpha 6
          Gene Aliases:
          E330019B14, E330026B02Rik
          Chromosome Location:
          Chr.9: 105689417 - 105828245 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01232629_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Col6a6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001102607.1 NP_001096077.1 16 - 17 4632 75
          GenBank mRNA AM748261.1 - 9 - 10 1448 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002997.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons

          Panther Classification:

          Molecular Function -

          extracellular matrix structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900024758.mm.1 NM_001102607.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024758.mm.1 NM_001102607.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024758.mm.1 TC0900002997.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900024761.mm.1 NM_001102607.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024761.mm.1 NM_001102607.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024761.mm.1 TC0900002997.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900024768.mm.1 NM_001102607.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024768.mm.1 NM_001102607.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024768.mm.1 TC0900002997.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900024753.mm.1 NM_001102607.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024753.mm.1 NM_001102607.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024753.mm.1 TC0900002997.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900024763.mm.1 NM_001102607.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024763.mm.1 NM_001102607.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024763.mm.1 TC0900002997.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900024764.mm.1 NM_001102607.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024764.mm.1 NM_001102607.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024764.mm.1 TC0900002997.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900024759.mm.1 NM_001102607.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024759.mm.1 NM_001102607.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024759.mm.1 TC0900002997.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900024762.mm.1 NM_001102607.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024762.mm.1 NM_001102607.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024762.mm.1 TC0900002997.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900024765.mm.1 NM_001102607.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024765.mm.1 NM_001102607.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024765.mm.1 TC0900002997.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900024760.mm.1 NM_001102607.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024760.mm.1 NM_001102607.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024760.mm.1 TC0900002997.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900024770.mm.1 NM_001102607.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024770.mm.1 NM_001102607.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024770.mm.1 TC0900002997.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900024757.mm.1 NM_001102607.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024757.mm.1 NM_001102607.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024757.mm.1 TC0900002997.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900024771.mm.1 NM_001102607.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024771.mm.1 NM_001102607.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024771.mm.1 TC0900002997.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900024766.mm.1 NM_001102607.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024766.mm.1 NM_001102607.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024766.mm.1 TC0900002997.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900024767.mm.1 NM_001102607.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024767.mm.1 NM_001102607.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024767.mm.1 TC0900002997.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900024769.mm.1 NM_001102607.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024769.mm.1 NM_001102607.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024769.mm.1 TC0900002997.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline