Search
Search

Tm9sf2| Gene Symbol: Tm9sf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080556.3 | NP_542123.3 | 7 - 8 | 1013 | 82 | |
| GenBank mRNA | AK010325.1 | - | 7 - 8 | 970 | 82 | |
| AK030340.1 | - | 7 - 8 | 944 | 82 | ||
| AK048012.1 | - | 7 - 8 | 932 | 82 | ||
| AK075720.1 | - | 7 - 8 | 1013 | 82 | ||
| AK088905.1 | - | 7 - 8 | 985 | 82 | ||
| AK159757.1 | - | 7 - 8 | 958 | 82 | ||
| AK212229.1 | - | 3 - 4 | 348 | 82 | ||
| BC003862.1 | - | 7 - 8 | 952 | 3488326 | 82 | |
Set Membership: |
|