Search
Search
| Gene Symbol: Cul2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029402.3 | NP_083678.2 | 18 - 19 | 2082 | 126 | |
| GenBank mRNA | AK004875.2 | - | 18 - 19 | 2038 | 126 | |
| AK028589.1 | - | 18 - 19 | 2051 | 126 | ||
| AK144693.1 | - | 18 - 19 | 2056 | 126 | ||
| AK160597.1 | - | 18 - 19 | 2067 | 126 | ||
| AK220287.1 | - | 18 - 19 | 2057 | 126 | ||
| BC025902.1 | - | 10 - 11 | 1148 | 126 | ||
| BC026779.1 | - | 18 - 19 | 1983 | 4018298 | 126 | |
| BC027428.1 | - | 18 - 19 | 2045 | 3983461 | 126 | |
Set Membership: |
|