Search
Search
| Gene Symbol: Epc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007935.2 | NP_031961.1 | 8 - 9 | 1325 | 73 | |
| NM_027497.3 | NP_081773.1 | 8 - 9 | 1476 | 73 | ||
| GenBank mRNA | AF079765.1 | - | 8 - 9 | 1304 | 73 | |
| AK040254.1 | - | 8 - 9 | 1476 | 73 | ||
| AK149317.1 | - | 5 - 6 | 676 | 73 | ||
| BC138622.1 | - | 8 - 9 | 1304 | 8861642 | 73 | |
| BC138623.1 | - | 8 - 9 | 1304 | 8861643 | 73 | |
Set Membership: |
|