Search
Search

Abcb7| Gene Symbol: Abcb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009592.1 | NP_033722.1 | 8 - 9 | 1046 | 145 | |
| GenBank mRNA | AK040069.1 | - | 8 - 9 | 1043 | 145 | |
| AK084853.1 | - | 8 - 9 | 1037 | 145 | ||
| AK151967.1 | - | 8 - 9 | 1032 | 145 | ||
| AK152535.1 | - | 8 - 9 | 1032 | 145 | ||
| AK152816.1 | - | 8 - 9 | 1032 | 145 | ||
| AK152946.1 | - | 8 - 9 | 1033 | 145 | ||
| BC035534.1 | - | 5 - 6 | 675 | 145 | ||
| BC151037.1 | - | 8 - 9 | 1046 | 9087950 | 145 | |
| U43892.1 | - | 7 - 8 | 850 | 145 | ||
Set Membership: |
|