Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01235278_m1
          See other 2610203C22RIK GE Assays ›
          Gene Symbol:
          2610203C22Rik
          Gene Name:
          RIKEN cDNA 2610203C22 gene
          Gene Aliases:
          2310007M24Rik
          Chromosome Location:
          Chr.1: 9560832 - 9631092 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01235278_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2610203C22Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015470.1 2 - 3 160 73
          GenBank mRNA AK009214.1 - 2 - 3 236 73
          AK011866.1 - 2 - 3 206 73
          BC037765.1 - 2 - 3 160 73
          BC064105.1 - 2 - 3 160 73
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY711084.1 - 2 - 3 206 Chr. 1 - 9563509 - 9631137 [-] on Build GRCm38 73
          GenBankmRNA AW908209.1 - 2 - 3 160 Chr. 1 - 9561667 - 9631092 [-] on Build GRCm38 73
          GenBankmRNA DV052199.1 - 270 Chr. - - [-] on Build 73
          GenBankmRNA CB590559.1 - 252 Chr. - - [-] on Build 73
          GenBankmRNA AW985729.1 - 2 - 3 160 Chr. 1 - 9561575 - 9631092 [-] on Build GRCm38 73
          GenBankmRNA BY709022.1 - 2 - 3 235 Chr. 1 - 9601023 - 9631162 [-] on Build GRCm38 73

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 3110035E14 gene 3110035E14Rik AI415019,
          AV164974,
          R75066
          Anti-sense Chr. 1 - 9601267 - 9627142 on NCBI Build GRCm38
          Downstream myeloblastosis oncogene-like 1 Mybl1 A-myb,
          G1-419-6,
          repro9
          46.3kb Sense Chr. 1 - 9667407 - 9700439 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001942.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100015909.mm.1 NR_015470.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015909.mm.1 NR_015470.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015909.mm.1 TC0100001942.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100015910.mm.1 NR_015470.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015910.mm.1 NR_015470.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015910.mm.1 TC0100001942.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100015911.mm.1 NR_015470.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015911.mm.1 NR_015470.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015911.mm.1 TC0100001942.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0