Search
Search

Thada| Gene Symbol: Thada | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183021.3 | NP_898842.2 | 25 - 26 | 3904 | 108 | |
| GenBank mRNA | AK173258.1 | - | 10 - 11 | 1414 | 108 | |
| EF222207.1 | - | 25 - 26 | 3812 | 108 | ||
Set Membership: |
|