Search
Search

Dalrd3| Gene Symbol: Dalrd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026378.3 | NP_080654.3 | 5 - 6 | 940 | 71 | |
| GenBank mRNA | AK149963.1 | - | 5 - 6 | 941 | 71 | |
| AK169405.1 | - | 5 - 6 | 938 | 71 | ||
| AK171818.1 | - | 5 - 6 | 941 | 71 | ||
| AK206691.1 | - | 3 - 4 | 287 | 71 | ||
| BC013483.1 | - | 5 - 6 | 921 | 4242135 | 71 | |
| BC058774.1 | - | 3 - 4 | 339 | 71 | ||
Set Membership: |
|