Search
Search

Acsbg2| Gene Symbol: Acsbg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039114.1 | NP_001034203.1 | 12 - 13 | 1975 | 66 | |
| GenBank mRNA | DQ250679.1 | - | 12 - 13 | 1975 | 66 | |
Set Membership: |
|