Search
Search
| Gene Symbol: Rfx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009056.2 | NP_033082.1 | 4 - 5 | 378 | 89 | |
| NM_027787.1 | NP_082063.1 | 4 - 5 | 378 | 89 | ||
| GenBank mRNA | AK044439.1 | - | 4 - 5 | 378 | 89 | |
| AK086559.1 | - | 4 - 5 | 310 | 89 | ||
| BC004654.1 | - | 4 - 5 | 395 | 3497910 | 89 | |
| X76089.1 | - | 4 - 5 | 333 | 89 | ||
Set Membership: |
|