Search
Search

Amdhd1| Gene Symbol: Amdhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027908.1 | NP_082184.1 | 1 - 2 | 201 | 105 | |
| GenBank mRNA | AK005066.1 | - | 1 - 2 | 157 | 105 | |
| AK090143.1 | - | 1 - 2 | 191 | 105 | ||
| BB856590.1 | - | 1 - 2 | 203 | 105 | ||
| BC022626.1 | - | 1 - 2 | 79 | 105 | ||
| BC043681.1 | - | 1 - 2 | 134 | 105 | ||
| BC055025.1 | - | 1 - 2 | 99 | 105 | ||
| BC118616.1 | - | 1 - 2 | 188 | 40107296 | 105 | |
| BC121826.1 | - | 1 - 2 | 187 | 40107295 | 105 | |
Set Membership: |
|