Search
Search

Tbc1d15| Gene Symbol: Tbc1d15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025706.3 | NP_079982.3 | 2 - 3 | 206 | 87 | |
| GenBank mRNA | AK010704.1 | - | 2 - 3 | 138 | 87 | |
| AK014477.1 | - | 2 - 3 | 138 | 87 | ||
| AK078875.1 | - | 2 - 3 | 152 | 87 | ||
| BB618866.1 | - | 2 - 3 | 210 | 87 | ||
| BC032087.1 | - | 2 - 3 | 134 | 87 | ||
| BC053395.1 | - | 2 - 3 | 187 | 30054233 | 87 | |
| CG784857.1 | - | 2 - 3 | 149 | 87 | ||
| DQ054831.1 | - | 2 - 3 | 137 | 87 | ||
Set Membership: |
|