Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01237804_g1
          See other BBS10 GE Assays ›
          Gene Symbol:
          Bbs10
          Gene Name:
          Bardet-Biedl syndrome 10 (human)
          Gene Aliases:
          1300007O09Rik, AI452285
          Chromosome Location:
          Chr.10: 111298679 - 111301736 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01237804_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bbs10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027914.1 NP_082190.1 1 - 2 224 90
          GenBank mRNA BB855633.1 - 1 - 2 225 90
          BY122358.1 - 1 - 2 194 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000001293.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          retina homeostasis
          nonmotile primary cilium assembly
          regulation of protein complex assembly
          photoreceptor cell maintenance
          chaperone-mediated protein complex assembly
          nucleotide binding
          RNA polymerase II repressing transcription factor binding
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1973543 AK004937.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1973543 NM_027914.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492742 AK004937.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492742 AK004937.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 404606 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 224788 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131941 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1973543 AK004937.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1973543 NM_027914.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492742 AK004937.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492742 AK004937.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492743 AK004937.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492742 AK004937.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492742 AK004937.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1430170_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1440787_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10366335 - -
          Mouse Genome 430 2.0 Array 1430170_at - -
          Mouse Genome 430 2.0 Array 1440787_PM_s_at - -
          Mouse Genome 430 2.0 Array 1430170_PM_at - -
          Mouse Genome 430 2.0 Array 1440787_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71769_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71769_at - -
          Affymetrix Mouse Clariom S Array PSR1000009663.mm.1 NM_027914.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000009663.mm.1 NM_027914.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000009663.mm.1 TC1000001293.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000009664.mm.1 NM_027914.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000009664.mm.1 NM_027914.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000009664.mm.1 TC1000001293.mm.1 MTA TC
          Mouse Expression Set 430 1430170_PM_at - -
          Mouse Expression Set 430 1440787_PM_s_at - -
          Mouse Expression Set 430 1430170_at - -
          Mouse Expression Set 430 1440787_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1430170_at - -
          Affymetrix HT Mouse Genome 430B Array 1430170_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440787_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1440787_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 337914 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1217075 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1238590 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline