Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01238643_s1
          See other TSIX GE Assays ›
          Gene Symbol:
          Tsix
          Gene Name:
          X (inactive)-specific transcript, opposite strand
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 103431517 - 103484957 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01238643_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price 434.00
          Your Price
          Online offer: 407.65
          407.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tsix
          Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002844.2
          4 2564 69
          GenBank mRNA AF138745.1 - LIT1604
          Tsix
          4 2559 69
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AJ421479.1 - Chr. - - [-] on Build 69

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream testis specific X-linked gene Tsx AV207184 -56.7kb Sense Chr. X - 103409101 - 103424588 on NCBI Build GRCm38
          Downstream Jpx transcript,
          Xist activator (non-protein coding)
          Jpx 2010000I03Rik,
          2510040I06Rik,
          Enox
          8.6kb Sense Chr. X - 103493576 - 103506425 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          dosage compensation by inactivation of X chromosome
          chromatin-mediated maintenance of transcription
          inactivation of paternal X chromosome
          protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2153116 AF138745.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 679577 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 615356 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 348689 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 59552 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 218687 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 22408 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 596996 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 198548 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 401085 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 319212 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 281417 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 907929 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923259 AF319547.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923259 AK010484.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923259 AK028274.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923259 AK031534.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923259 AK031679.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923259 AK147715.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923259 AK152989.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923259 AK155138.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923259 AK167603.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923259 AK168455.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923259 AK169255.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923259 AK170967.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923259 BC021900.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923259 BC043699.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923259 BC055026.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923259 BC062920.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923259 BC071251.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923259 NM_028041.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 453585 AK047974.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 453585 M30514.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 453585 NM_009604.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 453585 X03818.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 393052 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777265 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 93804 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 19989 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 940662 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 21422 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 151297 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 752059 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 930631 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 241942 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 796040 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 301114 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 924765 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 290563 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 565861 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 824458 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 603496 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 679202 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 381038 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 82221 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 88933 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 116573 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 26145 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 138497 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 17964 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147879 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P177061 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09380211 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09380080 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P177061 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09380179 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09380119 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03729505 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P147879 - -
          Whole Mouse Genome G4122A A_52_P245806 L04961.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P245806 NR_001463.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P245806 NR_001570.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P245806 AK039861.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P245806 NR_002844.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P177061 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03729505 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09380080 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P147879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32572879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32572840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32572834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14513324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14770008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14860037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32572884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32572830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32572883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14770007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32572877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32572875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32572833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32572878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14598430 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32572839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32572828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32572832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32572874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32572843 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1427596_PM_at - -
          Mouse Genome 430 2.0 Array 1427596_PM_at - -
          Mouse Genome 430 2.0 Array 1427596_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22097_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22097_at - -
          Affymetrix Mouse Clariom S Array PSR0X00006877.mm.1 NR_002844.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00006877.mm.1 NR_002844.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00006877.mm.1 TC0X00003398.mm.1 MTA TC
          Mouse Expression Set 430 1427596_at - -
          Mouse Expression Set 430 1427596_PM_at - -
          Mouse Genome 430A 2.0 Array 1427596_at - -
          Mouse Genome 430A 2.0 Array 1427596_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427596_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427596_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100730026 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256650 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline